(R)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride

Supplier: TCI America

Synonyms: (R)-(+)-5-Benzyl-2,2,3-trimethyl-4-imidazolidinone hydrochloride

T2990-1G T2990-250MG
TCT2990-1GEA 194.67 USD
TCT2990-1G TCT2990-250MG
(R)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride
(R)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one hydrochloride
Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD08276839
CAS Number: 323196-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR