2,2'-Thenil

Supplier: TCI America

Synonyms: 1,2-Di(thiophen-2-yl)ethane-1,2-dione

T2901-1G T2901-5G
TCT2901-1GEA 49.44 USD
TCT2901-1G TCT2901-5G
2,2'-Thenil
2,2'-Thenil
Formula: C₁₀H₆O₂S₂
MW: 222.29 g/mol
MDL Number: MFCD00173678
CAS Number: 7333-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR