3,4,5-Trifluorophenylboronic acid (Contains varying amounts of Anhydride)

Supplier: TCI America

T2300-25G T2300-5G
TCT2300-25GEA 86.52 USD
TCT2300-25G TCT2300-5G
3,4,5-Trifluorophenylboronic acid (Contains varying amounts of Anhydride)
3,4,5-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
MDL Number: MFCD02093069
CAS Number: 143418-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR