Triphenylsulfonium trifluoromethanesulfonate

Supplier: TCI America

T2180-1G T2180-5G
TCT2180-1GEA 67.98 USD
TCT2180-1G TCT2180-5G
Triphenylsulfonium trifluoromethanesulfonate
Triphenylsulfonium trifluoromethanesulfonate
Formula: C₁₉H₁₅F₃O₃S₂
MW: 412.44 g/mol
MDL Number: MFCD01940916
CAS Number: 66003-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR