Ro 5-3335 ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
7-Chloro-5-(1H-pyrrol-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one, 7-Chloro-5-(1H-pyrrol-2-yl)-1H-benzo[e][1,4]diazepin-2(3H)-one
R0223-10MG
R0223-50MG
TCR0223-10MGEA
190.55
USD
TCR0223-10MG
TCR0223-50MG
Ro 5-3335 ≥98.0% (by HPLC)
Ro 5-3335
Formula:
C₁₃H₁₀ClN₃O MW: 259.69 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00870704 CAS Number: 30195-30-3 |
Specification Test Results
Appearance | White to Light red to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...