Ru(OAc)2[(S)-xylbinap]
Supplier: TCI America
Synonyms:
Diacetato{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}ruthenium(II), Diacetato[(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl]ruthenium(II)
R0171-1G
R0171-200MG
TCR0171-1GEA
181.28
USD
TCR0171-1G
TCR0171-200MG
Ru(OAc)2[(S)-xylbinap]
Ru(OAc)2[(S)-xylbinap]
Formula:
C₅₆H₅₄O₄P₂Ru MW: 954.06 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09753022 CAS Number: 374067-49-9 |
Specification Test Results
Purity (GC) | min. 95.0 % |
Purity (HPLC) | min. 95.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...