Ethyl acetopyruvate

Supplier: TCI America

Synonyms: Ethyl 2,4-dioxopentanoate, Ethyl 2,4-dioxovalerate

R0165-25G R0165-5G
TCR0165-25GEA 195.7 USD
TCR0165-25G TCR0165-5G
Ethyl acetopyruvate
Ethyl acetopyruvate
Formula: C₇H₁₀O₄
MDL Number: MFCD00009124
CAS Number: 615-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR