6-Quinolineboronic acid (Contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Quinolin-6-ylboronic acid, Quinoline-6-boronic acid

Q0098-1G Q0098-5G
TCQ0098-1GEA 52.53 USD
TCQ0098-1G TCQ0098-5G
6-Quinolineboronic acid (Contains varying amounts of Anhydride)
6-Quinolineboronic acid
Formula: C₉H₈BNO₂
MW: 172.98 g/mol
MDL Number: MFCD03093100
CAS Number: 376581-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR