Potassium-2,3,3-trimethyl-3H-indole-5-sulfonate

Supplier: TCI America

P2824-1G P2824-5G
TCP2824-1GEA 48.41 USD
TCP2824-1G TCP2824-5G
Potassium-2,3,3-trimethyl-3H-indole-5-sulfonate
Potassium-2,3,3-trimethyl-3H-indole-5-sulfonate
Formula: C₁₁H₁₂KNO₃S
MDL Number: MFCD07369684
CAS Number: 184351-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR