1-Phenyl-1H-imidazole trifluoromethanesulfonate
Supplier: TCI America
Synonyms:
1-Phenyl-1H-imidazol-3-ium trifluoromethanesulphonate
P2822-25G
P2822-5G
TCP2822-25GEA
656.11
USD
TCP2822-25G
TCP2822-5G
1-Phenyl-1H-imidazole trifluoromethanesulfonate
1-Phenyl-1H-imidazole trifluoromethanesulfonate
Formula:
C₁₀H₉F₃N₂O₃S |
MDL Number:
MFCD32206610 CAS Number: 361447-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...