3-(3-Pyridyl)-D-alanine dihydrochloride

Supplier: TCI America

P2805-1G P2805-250MG
TCP2805-1GEA 241.02 USD
TCP2805-1G TCP2805-250MG
3-(3-Pyridyl)-D-alanine dihydrochloride
3-(3-Pyridyl)-D-alanine dihydrochloride
Formula: C₈H₁₀N₂O₂·2ClH
MDL Number: MFCD00191179
CAS Number: 93960-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR