4-(Benzyloxy)benzenesulfonyl chloride

Supplier: TCI America

P2803-1G P2803-5G
TCP2803-1GEA 83.43 USD
TCP2803-1G TCP2803-5G
4-(Benzyloxy)benzenesulfonyl chloride
4-(Benzyloxy)benzenesulfonyl chloride
Formula: C₁₃H₁₁ClO₃S
MW: 282.75 g/mol
MDL Number: MFCD04035557
CAS Number: 87001-32-9
UN: 3261

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR