4-(Pyridin-4-yl)aniline
Supplier: TCI America
P2782-1G
P2782-5G
TCP2782-1GEA
163.77
USD
TCP2782-1G
TCP2782-5G
4-(Pyridin-4-yl)aniline
4-(Pyridin-4-yl)aniline
Formula:
C₁₁H₁₀N₂ MW: 170.2105 g/mol |
MDL Number:
MFCD00970053 CAS Number: 13296-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...