Pimobendan

Supplier: TCI America

P2779-250MG P2779-50MG
TCP2779-250MGEA 271.92 USD
TCP2779-250MG TCP2779-50MG
Pimobendan
Pimobendan
Formula: C₁₉H₁₈N₄O₂
MDL Number: MFCD00761648
CAS Number: 74150-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR