L-β-Homoproline hydrochloride

Supplier: TCI America

Synonyms: (S)-2-(Pyrrolidin-2-yl)acetic acid hydrochloride, H-β-HoPro-OH·HCl

P2777-1G P2777-250MG
TCP2777-1GEA 232.78 USD
TCP2777-1G TCP2777-250MG
L-β-Homoproline hydrochloride
L-beta-Homoproline hydrochloride
Formula: C₆H₁₂ClNO₂
MW: 165.617 g/mol
MDL Number: MFCD07363484
CAS Number: 53912-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR