(R)-4-Chloro-N-(quinuclidin-3-yl)benzamide

Supplier: TCI America

Synonyms: PNU-282987 free base

P2774-10MG P2774-50MG
TCP2774-10MGEA 123.6 USD
TCP2774-10MG TCP2774-50MG
(R)-4-Chloro-N-(quinuclidin-3-yl)benzamide
(R)-4-Chloro-N-(quinuclidin-3-yl)benzamide
Formula: C₁₄H₁₇ClN₂O
MW: 264.75 g/mol
MDL Number: MFCD08703110
CAS Number: 711085-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR