2,2'-[1,4-Phenylenebis(oxy-2,1-ethanediyloxy)]diethanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,4-Bis(1,4,7-trioxaheptyl)benzene, 1,4-Bis[2-(2-hydroxyethoxy)ethoxy]benzene, 2-[2-[4-[2-(2-Hydroxyethoxy)ethoxy]phenoxy]ethoxy]ethanol

P2762-1G P2762-5G
TCP2762-1GEA 130.81 USD
TCP2762-1G TCP2762-5G
2,2'-[1,4-Phenylenebis(oxy-2,1-ethanediyloxy)]diethanol ≥98.0% (by GC)
2,2'-[1,4-Phenylenebis(oxy-2,1-ethanediyloxy)]diethanol
Formula: C₁₄H₂₂O₆
MW: 286.32 g/mol
MDL Number: MFCD01109251
CAS Number: 35648-87-4

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR