Potassium tetraphenylborate

Supplier: TCI America

P2661-1G P2661-5G
TCP2661-1GEA 87.55 USD
TCP2661-1G TCP2661-5G
Potassium tetraphenylborate
Potassium tetraphenylborate
Formula: C₂₄H₂₀BK
MDL Number: MFCD03093877
CAS Number: 3244-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR