Potassium (4-acetylphenyl)trifluoroborate

Supplier: TCI America

Synonyms: Potassium 4-acetylphenyltrifluoroborate, Potassium (4-acetylphenyl)trifluoroboranuide

P2652-1G P2652-5G
TCP2652-1GEA 72.1 USD
TCP2652-1G TCP2652-5G
Potassium (4-acetylphenyl)trifluoroborate
Potassium 4-acetylphenyltrifluoroborate
Formula: C₈H₇BF₃KO
MW: 226.05 g/mol
MDL Number: MFCD07784376
CAS Number: 252726-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR