Pseudopelletierine

Supplier: TCI America

Synonyms: 9-Methyl-9-azabicyclo[3.3.1]nonan-3-one

P2648-1G P2648-5G
TCP2648-1GEA 52.53 USD
TCP2648-1G TCP2648-5G
Pseudopelletierine
Pseudopelletierine
Formula: C₉H₁₅NO
MW: 153.23 g/mol
MDL Number: MFCD00078161
CAS Number: 552-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR