Potassium(4-bromophenyl)trifluoroborate

Supplier: TCI America

Synonyms: (4-Bromophenyl)trifluoroboric acid potassium salt, Potassium (4-bromophenyl)trifluoroborate, Potassium 4-bromophenyltrifluoroborate

P2647-1G P2647-5G
TCP2647-1GEA 32.96 USD
TCP2647-1G TCP2647-5G
Potassium(4-bromophenyl)trifluoroborate
Potassium(4-bromophenyl)trifluoroborate
Formula: C₆H₄BBrF₃K
MW: 262.91 g/mol
MDL Number: MFCD02093978
CAS Number: 374564-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR