4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate

Supplier: TCI America

P2633-25G P2633-5G
TCP2633-25GEA 189.52 USD
TCP2633-25G TCP2633-5G
4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate
4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate
Formula: C₃₁H₄₈O₂
MDL Number: MFCD00673705
CAS Number: 88038-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR