6-Azaindole

Supplier: TCI America

P2530-1G P2530-5G
TCP2530-1GEA 44.29 USD
TCP2530-1G TCP2530-5G
6-Azaindole
6-Azaindole
Formula: C₇H₆N₂
MDL Number: MFCD01686899
CAS Number: 271-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR