Picolinic acid N-oxide

Supplier: TCI America

P2516-25G P2516-5G
TCP2516-25GEA 318.27 USD
TCP2516-25G TCP2516-5G
Picolinic acid N-oxide
Picolinic acid N-oxide
Formula: C₆H₅NO₃
MW: 139.1088 g/mol
MDL Number: MFCD00006198
CAS Number: 824-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR