4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide

Supplier: TCI America

Synonyms: Prucalopride

P2467-10MG P2467-50MG
TCP2467-10MGEA 63.86 USD
TCP2467-10MG TCP2467-50MG
4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide
4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide
Formula: C₁₈H₂₆ClN₃O₃
MW: 367.87 g/mol
MDL Number: MFCD09837787
CAS Number: 179474-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR