2-Phenoxybenzhydrazide ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P2357-1G P2357-5G
TCP2357-1GEA 60.77 USD
TCP2357-1G TCP2357-5G
2-Phenoxybenzhydrazide ≥98.0% (by GC, titration analysis)
2-Phenoxybenzhydrazide
Formula: C₁₃H₁₂N₂O₂
MDL Number: MFCD00017048
CAS Number: 43038-37-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 101.0 to 105.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR