Palmatine chloride

Supplier: TCI America

Synonyms: 2,3,9,10-Tetramethoxy-5,6-dihydroisoquinolino[3,2-a]isoquinolin-7-ium chloride

P2138-1G P2138-5G
TCP2138-1GEA 119.48 USD
TCP2138-1G TCP2138-5G
Palmatine chloride
Palmatine chloride
Formula: C₂₁H₂₂ClNO₄
MDL Number: MFCD00016656
CAS Number: 10605-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR