(2-Aminoethoxy)benzene

Supplier: TCI America

Synonyms: ω-Aminophenetole, 2-Phenoxyethylamine, 2-Aminoethyl phenyl ether, β-Aminophenetole

P1663-25G P1663-5G
TCP1663-25GEA 215.27 USD
TCP1663-25G TCP1663-5G
(2-Aminoethoxy)benzene
(2-Aminoethoxy)benzene
Formula: C₆H₅OCH₂CH₂NH₂
MW: 137.18 g/mol
MDL Number: MFCD00052975
CAS Number: 1758-46-9
UN: 2735

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR