Potassium 2-phenylethyltrifluoroborate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Potassium Trifluoro(phenethyl)borate, Potassium Trifluoro(2-phenylethyl)borate, Potassium Phenethyltrifluoroborate

P1581-1G P1581-5G
TCP1581-1GEA 54.59 USD
TCP1581-1G TCP1581-5G
Potassium 2-phenylethyltrifluoroborate ≥98.0% (by HPLC)
Potassium 2-phenylethyltrifluoroborate
Formula: C₈H₉BF₃K
Melting Pt: 312°C
MDL Number: MFCD09039257
CAS Number: 329976-74-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR