Potassium methyltrifluoroborate ≥97.0% (W)

Supplier: TCI America
Warning

Synonyms: Potassium trifluoro(methyl)borate, Methyltrifluoroboric acid potassium salt

P1478-1G P1478-5G
TCP1478-1GEA 16.48 USD
TCP1478-1G TCP1478-5G
Potassium methyltrifluoroborate ≥97.0% (W)
Potassium methyltrifluoroborate
Formula: CH₃BF₃K
MDL Number: MFCD08276757
CAS Number: 13862-28-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Gravimetric Method) min. 97.0 %
Melting point 287.0 to 291.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR