3-Oxo-3-(2-thienyl)propionitrile

Supplier: TCI America

O0571-25G O0571-5G
TCO0571-25GEA 258.53 USD
TCO0571-25G TCO0571-5G
3-Oxo-3-(2-thienyl)propionitrile
3-Oxo-3-(2-thienyl)propionitrile
Formula: C₇H₅NOS
MW: 151.19 g/mol
MDL Number: MFCD00052586
CAS Number: 33898-90-7
UN: 3439

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR