Oxybutynin hydrochloride

Supplier: TCI America

O0451-1G O0451-5G
TCO0451-1GEA 144.2 USD
TCO0451-1G TCO0451-5G
Oxybutynin hydrochloride
Oxybutynin hydrochloride
Formula: C₂₂H₃₁NO₃·HCl
MDL Number: MFCD00072150
CAS Number: 1508-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR