Oleuropein

Supplier: TCI America

Synonyms: Methyl (2S,4S,E)-4-(2-(3,4-dihydroxyphenethoxy)-2-oxoethyl)-3-ethylidene-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydro-2H-pyran-5-carboxylate

O0420-100MG O0420-25MG
TCO0420-100MGEA 217.33 USD
TCO0420-100MG TCO0420-25MG
Oleuropein
Oleuropein
Formula: C₂₅H₃₂O₁₃
MW: 540.51 g/mol
MDL Number: MFCD16879008
CAS Number: 32619-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR