2,3,5-Trimethyl-4-nitropyridine N-Oxide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2,3,5-Trimethyl-4-nitropyridine 1-Oxide
N1189-1G
N1189-250MG
TCN1189-1GEA
167.89
USD
TCN1189-1G
TCN1189-250MG
2,3,5-Trimethyl-4-nitropyridine N-Oxide ≥98.0% (by GC, titration analysis)
2,3,5-Trimethyl-4-nitropyridine N-Oxide
Formula:
C₈H₁₀N₂O₃ MW: 182.18 g/mol |
MDL Number:
MFCD00800254 CAS Number: 86604-79-7 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...