Nitrendipine
Supplier: TCI America
Synonyms:
3-Ethyl 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
N1186-1G
N1186-250MG
TCN1186-1GEA
191.58
USD
TCN1186-1G
TCN1186-250MG
Nitrendipine
Nitrendipine
Formula:
C₁₈H₂₀N₂O₆ MW: 360.366 g/mol |
MDL Number:
MFCD00082255 CAS Number: 39562-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...