Nifuroxazide

Supplier: TCI America

N1181-100MG N1181-500MG N1181-5G
TCN1181-100MGEA 35.02 USD
TCN1181-100MG TCN1181-500MG TCN1181-5G
Nifuroxazide
Nifuroxazide
Formula: C₁₂H₉N₃O₅
MDL Number: MFCD00079482
CAS Number: 965-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR