3-Nitrophenyl isothiocyanate

Supplier: TCI America

Synonyms: 1-Isothiocyanato-3-nitrobenzene, m-Nitrophenylisothiocyanat

N1180-5G N1180-25G
TCN1180-5GEA 84.46 USD
TCN1180-5G TCN1180-25G
3-Nitrophenyl isothiocyanate
3-Nitrophenyl isothiocyanate
Formula: C₇H₄N₂O₂S
MDL Number: MFCD00041245
CAS Number: 3529-82-6
UN: 2811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR