1H,1H,2H,2H-Nonafluorohexyl methacrylate stabilised

Supplier: TCI America

Synonyms: Methacrylic acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl methacrylate, Methacrylic acid 1H,1H,2H,2H-Nonafluorohexyl ester

Stabilization: Stabilised with TBC

N1143-25G N1143-5G
TCN1143-25GEA 162.74 USD
TCN1143-25G TCN1143-5G
1H,1H,2H,2H-Nonafluorohexyl methacrylate stabilised
1H,1H,2H,2H-Nonafluorohexyl methacrylate
Formula: C₁₀H₉F₉O₂
MW: 332.17 g/mol
MDL Number: MFCD00236094
CAS Number: 1799-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR