Nickel(II) bromide 2-methoxyethyl ether complex

Supplier: TCI America

N1049-10G N1049-1G
TCN1049-10GEA 96.82 USD
TCN1049-10G TCN1049-1G
Nickel(II) bromide 2-methoxyethyl ether complex
Nickel(II) bromide 2-methoxyethyl ether complex
Formula: C₆H₁₄Br₂NiO₃
MW: 352.6700 g/mol
MDL Number: MFCD00274339
CAS Number: 312696-09-6
UN: 1325

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR