2-Methoxy-5-nitrobenzaldehyde

Supplier: TCI America

N1034-25G N1034-5G
TCN1034-25GEA 111.24 USD
TCN1034-25G TCN1034-5G
2-Methoxy-5-nitrobenzaldehyde
2-Methoxy-5-nitrobenzaldehyde
Formula: C₈H₇NO₄
MDL Number: MFCD02093683
CAS Number: 25016-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR