4-Nitrophenyl phenyl ether

Supplier: TCI America

N1011-25G N1011-5G
TCN1011-25GEA 128.75 USD
TCN1011-25G TCN1011-5G
4-Nitrophenyl phenyl ether
4-Nitrophenyl phenyl ether
Formula: C₁₂H₉NO₃
MDL Number: MFCD00007320
CAS Number: 620-88-2
UN: 3077

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR