4-Methoxybutanoic acid

Supplier: TCI America

M3329-1G M3329-250MG
TCM3329-1GEA 197.76 USD
TCM3329-1G TCM3329-250MG
4-Methoxybutanoic acid
4-Methoxybutanoic acid
Formula: C₅H₁₀O₃
MW: 118.131 g/mol
MDL Number: MFCD00029855
CAS Number: 29006-02-8
UN: 3265

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR