Methyl 5-formyl-2-methoxybenzoate

Supplier: TCI America

M3327-25G M3327-5G
TCM3327-25GEA 339.9 USD
TCM3327-25G TCM3327-5G
Methyl 5-formyl-2-methoxybenzoate
Methyl 5-formyl-2-methoxybenzoate
Formula: C₁₀H₁₀O₄
MDL Number: MFCD10565654
CAS Number: 78515-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR