3-Methyl-4-nitroquinoline 1-Oxide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Methyl-4-nitroquinoline N-Oxide

M3311-1G M3311-250MG
TCM3311-1GEA 266.77 USD
TCM3311-1G TCM3311-250MG
3-Methyl-4-nitroquinoline 1-Oxide ≥98.0% (by HPLC, titration analysis)
3-Methyl-4-nitroquinoline 1-Oxide
Formula: C₁₀H₈N₂O₃
MW: 204.19 g/mol
MDL Number: MFCD01685462
CAS Number: 14073-00-8

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR