Methyl 4-Isopropylbenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Isopropylbenzoic acid methyl ester, Methyl 4-isopropylbenzoate, Methyl Cumate

M3307-1G M3307-5G
TCM3307-1GEA 49.44 USD
TCM3307-1G TCM3307-5G
Methyl 4-Isopropylbenzoate ≥98.0% (by GC)
Methyl 4-Isopropylbenzoate
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 126°C
MDL Number: MFCD06203666
CAS Number: 20185-55-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR