N-Methoxycarbonyl-D-phenylglycine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

M3293-100G M3293-25G
TCM3293-100GEA 74.16 USD
TCM3293-100G TCM3293-25G
N-Methoxycarbonyl-D-phenylglycine ≥97.0% (by HPLC, titration analysis)
N-Methoxycarbonyl-D-phenylglycine
Formula: C₁₀H₁₁NO₄
MDL Number: MFCD12796004
CAS Number: 50890-96-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 117.0 to 121.0 deg-C
Specific rotation [a]20/D +3.0 to +5.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR