4'-Methoxy-2-chloroacetophenone

Supplier: TCI America

Synonyms: 2-Chloro-1-(4-methoxyphenyl)ethanone, 4-Methoxyphenacyl chloride

M3268-25G M3268-5G
TCM3268-25GEA 389.34 USD
TCM3268-25G TCM3268-5G
4'-Methoxy-2-chloroacetophenone
4'-Methoxy-2-chloroacetophenone
Formula: C₉H₉ClO₂
MW: 184.6196 g/mol
MDL Number: MFCD00216508
CAS Number: 2196-99-8
UN: 3261

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR