2-Methoxy-5-nitropyrimidine

Supplier: TCI America

M3080-1G M3080-5G
TCM3080-1GEA 124.63 USD
TCM3080-1G TCM3080-5G
2-Methoxy-5-nitropyrimidine
2-Methoxy-5-nitropyrimidine
Formula: C₅H₅N₃O₃
MDL Number: MFCD00474639
CAS Number: 14001-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR