Metamizole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methylamino)methanesulphonic acid sodium salt, Sulpyrine, Sodium (1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methylamino)methanesulphonate, Metamizole sodium salt, Metamizole (sodium salt), Dipyrone

M3060-10G M3060-50G
TCM3060-10GEA 62.83 USD
TCM3060-10G TCM3060-50G
Metamizole ≥98.0% (by HPLC, titration analysis)
Metamizole
Formula: C₁₃H₁₆N₃O₄S·Na
Melting Pt: 227°C
MDL Number: MFCD00020783
CAS Number: 68-89-3
EINECS: 200-694-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR