Methyl tosylcarbamate

Supplier: TCI America

M2988-10G M2988-1G
TCM2988-10GEA 659.2 USD
TCM2988-10G TCM2988-1G
Methyl tosylcarbamate
Methyl tosylcarbamate
Formula: C₉H₁₁NO₄S
MDL Number: MFCD00453717
CAS Number: 14437-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR